Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172584.3 | NP_766172.1 | 10 | 971 | ||
GenBank mRNA | AK038913.1 | - | 10 | 972 | ||
BC025917.1 | - | 3 | 251 | |||
BC031182.1 | - | 2 | 33 | |||
BC056464.1 | - | 10 | 952 | 5707377 |