Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 23 | 2583 | ||
NM_001291444.1 | NP_001278373.1 | 23 | 2589 | |||
NM_001291445.1 | NP_001278374.1 | 22 | 2493 | |||
GenBank mRNA | AK030003.1 | - | 18 | 1952 | ||
AK036581.1 | - | 22 | 2469 | |||
AK054000.1 | - | 23 | 2564 | |||
AK122326.1 | - | 21 | 2272 | |||
AK193976.1 | - | 2 | 310 | |||
BC043302.1 | - | 4 | 500 | |||
BC059076.1 | - | 8 | 1153 |