Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160385.2 | NP_001153857.1 | 2, 3 | 716 | ||
NM_001294146.1 | NP_001281075.1 | 2, 3 | 734 | |||
NM_001294154.1 | NP_001281083.1 | 2, 3 | 716 | |||
GenBank mRNA | BB626213.1 | - | 2, 3 | 120 | ||
BU604736.1 | - | 2, 3 | 288 |