Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281496.1 | NP_001268425.1 | 3 | 330 | ||
NM_001281497.1 | NP_001268426.1 | 3 | 282 | |||
NM_001281715.1 | NP_001268644.1 | 4 | 374 | |||
NM_002634.3 | NP_002625.1 | 3 | 282 | |||
XM_017024762.1 | XP_016880251.1 | 3 | 254 | |||
XM_017024763.1 | XP_016880252.1 | 4 | 424 | |||
GenBank mRNA | AK223181.1 | - | 3 | 257 | ||
AK290766.1 | - | 3 | 257 | |||
AK302258.1 | - | 3 | 257 | |||
AK307767.1 | - | 3 | 254 | |||
AK312649.1 | - | 3 | 257 | |||
BC013401.2 | - | 3 | 242 | 3010198 | ||
BC095460.1 | - | 3 | 231 | 30557359 | ||
BG029858.1 | - | 4 | 335 | |||
BT007411.1 | - | 2 | 184 | |||
CR541706.1 | - | 2 | 184 | |||
DC360133.1 | - | 3 | 330 | |||
S85655.1 | - | 3 | 234 |