Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172590.3 | NP_766178.2 | 9 | 827 | ||
GenBank mRNA | AK046853.1 | - | 9 | 796 | ||
AK082348.1 | - | 9 | 828 | |||
AK149993.1 | - | 9 | 793 | |||
BC138095.1 | - | 9 | 733 | 8861114 | ||
BC145143.1 | - | 9 | 729 | |||
BC145144.1 | - | 9 | 733 | 9053695 |