Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172591.3 | NP_766179.2 | 4 | 557 | ||
GenBank mRNA | AK031041.1 | - | 4 | 560 | ||
AK142282.1 | - | 4 | 243 | |||
AK146566.1 | - | 4 | 291 | |||
AK164135.1 | - | 4 | 303 | |||
BB620239.1 | - | 4 | 560 | |||
BC019933.1 | - | 5 | 293 | |||
BC047122.1 | - | 5 | 293 | |||
BC052456.1 | - | 4 | 254 | 6830607 | ||
BC053718.1 | - | 4 | 273 |