Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289760.1 | NP_001276689.1 | 8 | 1431 | ||
NM_001289761.1 | NP_001276690.1 | 7 | 1350 | |||
NM_001289762.1 | NP_001276691.1 | 6 | 960 | |||
NM_011243.2 | NP_035373.1 | 6 | 1314 | |||
GenBank mRNA | AK078471.1 | - | 6 | 1026 | ||
BC076597.1 | - | 6 | 1310 | 30608242 | ||
S56660.1 | - | 6 | 1210 |