Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145462.2 | NP_663437.2 | 10 | 1176 | ||
GenBank mRNA | AK079161.1 | - | 10 | 1116 | ||
AK079852.1 | - | 10 | 1144 | |||
AK087969.1 | - | 10 | 1149 | |||
AK207390.1 | - | 1 | 40 | |||
AK219765.1 | - | 1 | 40 | |||
BC004644.1 | - | 10 | 1151 | 3157901 | ||
BC011095.1 | - | 10 | 1151 | 3157901 |