Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253752.1 | NP_001240681.1 | 7 | 1358 | ||
NM_001253753.1 | NP_001240682.1 | 7 | 1428 | |||
NM_144842.3 | NP_659091.3 | 7 | 1422 | |||
GenBank mRNA | AK036513.1 | - | 6 | 1267 | ||
AK155333.1 | - | 7 | 1340 | |||
AK156501.1 | - | 7 | 1430 | |||
BC027380.1 | - | 5 | 771 | |||
CN534727.1 | - | 1 | 139 |