Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145469.5 | NP_663444.2 | 2, 3 | 212 | ||
GenBank mRNA | AK033975.1 | - | 2, 3 | 213 | ||
AK038556.1 | - | 2, 3 | 353 | |||
AK079085.1 | - | 2, 3 | 645 | |||
AK132530.1 | - | 2, 3 | 240 | |||
BC016107.1 | - | 2, 3 | 235 | 4921508 | ||
BC030399.1 | - | 2, 3 | 729 | 4166825 | ||
BC058207.1 | - | 2, 3 | 378 | 5361417 |