Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E430025E21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153548.2 | NP_705776.2 | 24, 25 | 3180 | ||
GenBank mRNA | AK088754.1 | - | 9, 10 | 1053 | ||
AK128970.1 | - | 24, 25 | 3195 | |||
AK129086.1 | - | 2832 | ||||
AK163968.1 | - | 7, 8 | 804 | |||
BC031364.1 | - | 10, 11 | 1194 | |||
BC034070.1 | - | 14, 15 | 1780 | 4217923 | ||
BC040815.1 | - | 14, 15 | 1780 | 4217923 | ||
BC067035.1 | - | 24, 25 | 3180 | 30532658 |