Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145474.2 | NP_663449.2 | 2 | 379 | ||
GenBank mRNA | BC018285.1 | - | 2 | 379 | 4207630 |