Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030014.2 | NP_001025185.1 | 4 | 876 | ||
GenBank mRNA | AK135457.1 | - | 4 | 902 | ||
BC025519.1 | - | 2 | 372 | |||
BC099494.1 | - | 4 | 876 | 30844555 |