Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083114.1 | NP_001076583.1 | 4 | 311 | ||
NM_001285863.1 | NP_001272792.1 | 4 | 440 | |||
NM_001285864.1 | NP_001272793.1 | 3 | 219 | |||
NM_146062.3 | NP_666174.2 | 5 | 518 | |||
NM_175363.4 | NP_780572.1 | 4 | 311 | |||
GenBank mRNA | AK081606.1 | - | 4 | 304 | ||
AK150904.1 | - | 4 | 262 | |||
AK158488.1 | - | 4 | 441 | |||
AK160138.1 | - | 4 | 284 | |||
AK168443.1 | - | 4 | 312 | |||
BC031486.1 | - | 5 | 471 | 3157250 | ||
BC150854.2 | - | 3 | 215 | 9087765 | ||
BG145871.1 | - | 5 | 440 | |||
BY006733.1 | - | 4 | 312 | |||
CF734096.1 | - | 3 | 199 |