Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spidr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146068.4 | NP_666180.2 | 6 | 735 | ||
GenBank mRNA | AK044836.1 | - | 6 | 718 | ||
AK052832.1 | - | 6 | 698 | |||
AK075871.1 | - | 6 | 700 | |||
AK146053.1 | - | 6 | 735 | |||
AK166448.1 | - | 2 | 131 | |||
AK170784.1 | - | 2 | 367 | |||
AK172902.1 | - | 6 | 688 | |||
BC089026.1 | - | 6 | 672 | 30615298 |