Gene Symbol: Hectd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163471.1 | NP_001156943.1 | 8 | 898 | ||
NM_172637.3 | NP_766225.2 | 8 | 898 | |||
GenBank mRNA | AK029549.1 | - | 8 | 758 | ||
AK043646.1 | - | 8 | 859 | |||
AK142777.1 | - | 6 | 713 | |||
BC137671.1 | - | 8 | 869 | 8860690 | ||
BC144815.1 | - | 8 | 869 | 9053363 | ||
CB526486.1 | - | 5 | 351 |