Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130525.1 | NP_001123997.1 | 2 | 238 | ||
NM_001130526.1 | NP_001123998.1 | 2 | 230 | |||
NM_145503.2 | NP_663478.2 | 2 | 282 | |||
GenBank mRNA | AK081367.1 | - | 2 | 154 | ||
AK129446.1 | - | 1 | 147 | |||
AK144154.1 | - | 2 | 249 | |||
AK169936.1 | - | 2 | 271 | |||
AK203886.1 | - | 1 | 50 | |||
BC014695.1 | - | 1 | 131 | 3157493 | ||
BC092263.1 | - | 2 | 262 | 2654420 | ||
BX511791.1 | - | 2 | 238 | |||
BY202542.1 | - | 2 | 283 | |||
BY726859.1 | - | 2 | 248 | |||
CK021048.1 | - | 2 | 239 |