Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005028.4 | NP_005019.2 | 4 | 659 | ||
XM_011519496.1 | XP_011517798.1 | 4 | 435 | |||
XM_017016330.1 | XP_016871819.1 | 4 | 493 | |||
XM_017016331.1 | XP_016871820.1 | 4 | 588 | |||
GenBank mRNA | AK294817.1 | - | 4 | 440 | ||
BC018034.1 | - | 4 | 640 | 4817357 | ||
S78798.1 | - | 4 | 725 | |||
U14957.2 | - | 4 | 433 |