Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kansl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172652.3 | NP_766240.1 | 7 | 944 | ||
GenBank mRNA | AK030079.1 | - | 7 | 945 | ||
AK129329.1 | - | 7 | 808 | |||
BC132293.1 | - | 6 | 847 | 40130570 | ||
BC144763.1 | - | 6 | 866 |