Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B3gnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145222.2 | NP_660257.2 | 2 | 913 | ||
GenBank mRNA | AF502429.1 | - | 2 | 890 | ||
AK049105.1 | - | 2 | 1298 | |||
AK080081.1 | - | 2 | 901 | |||
AK131975.1 | - | 2 | 704 | |||
AK133674.1 | - | 2 | 652 | |||
AK154691.1 | - | 2 | 665 | |||
BC031187.1 | - | 2 | 666 | 4990205 | ||
BC060507.1 | - | 2 | 632 | 6394682 |