Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144886.2 | NP_659135.1 | 7 | 622 | ||
GenBank mRNA | AK144234.1 | - | 7 | 631 | ||
AK164618.1 | - | 6 | 1404 | |||
AK166881.1 | - | 7 | 632 | |||
BC021485.1 | - | 7 | 646 | 5344620 | ||
BC021807.1 | - | 7 | 644 | 3980155 |