Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 12 | 1473 | ||
GenBank mRNA | BC046474.1 | - | 12 | 1579 | ||
CJ235603.1 | - | 6 | 404 | |||
HQ426680.1 | - | 12 | 1473 | |||
HQ426681.1 | - | 10 | 1266 | |||
HQ426682.1 | - | 9 | 1134 | |||
HQ426683.1 | - | 8 | 891 | |||
HQ426684.1 | - | 11 | 1341 | |||
HQ426685.1 | - | 11 | 1413 | |||
HQ426686.1 | - | 11 | 1330 | |||
HQ426687.1 | - | 12 | 1604 | |||
HQ426688.1 | - | 12 | 1620 | |||
HQ426689.1 | - | 11 | 1488 | |||
HQ426690.1 | - | 12 | 1690 | |||
HQ426691.1 | - | 1272 | ||||
HQ426692.1 | - | 1403 | ||||
HQ426693.1 | - | 1313 | ||||
HQ426694.1 | - | 1226 | ||||
HQ426695.1 | - | 1445 | ||||
HQ426696.1 | - | 1385 | ||||
HQ426697.1 | - | 1530 | ||||
HQ426698.1 | - | 1238 | ||||
HQ426699.1 | - | 1302 | ||||
HQ426700.1 | - | 1576 | ||||
HQ426701.1 | - | 1592 | ||||
HQ426702.1 | - | 11 | 1055 | |||
HQ426703.1 | - | 9 | 848 | |||
HQ426704.1 | - | 12 | 1212 | |||
HQ426705.1 | - | 10 | 912 | |||
HQ426706.1 | - | 10 | 999 | |||
HQ426707.1 | - | 10 | 1146 | |||
HQ426708.1 | - | 6 | 509 | |||
HQ426709.1 | - | 9 | 904 | |||
HQ426710.1 | - | 8 | 844 | |||
HQ426711.1 | - | 9 | 1051 |