Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207236.1 | NP_997119.1 | 1 | 564 | ||
GenBank mRNA | BC125536.1 | - | 1 | 607 | 40130051 |
Gene Symbol: Olfr1280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146908.1 | NP_667119.1 | 1 | 564 | ||
GenBank mRNA | BC119051.1 | - | 1 | 607 | 8733800 | |
BC119053.1 | - | 1 | 607 | 8733802 |
Gene Symbol: Olfr1281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005568.1 | NP_001005568.1 | 1 | 564 |
Gene Symbol: Olfr1282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146907.2 | NP_667118.2 | 1 | 564 |
Gene Symbol: Olfr1286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207254.1 | NP_997137.1 | 1 | 564 |
Gene Symbol: Olfr1287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011773.1 | NP_001011773.1 | 1 | 564 |