Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Csrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166640.1 | NP_001160112.1 | 9 | 1975 | ||
NM_181417.3 | NP_852082.2 | 9 | 2458 | |||
GenBank mRNA | AK088564.1 | - | 1 | 178 | ||
AK191012.1 | - | 2 | 240 | |||
BC031563.1 | - | 9 | 2458 | 4949236 | ||
BC033464.1 | - | 9 | 1975 |