Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199304.1 | NP_955008.2 | 9 | 1231 | ||
GenBank mRNA | AK089176.1 | - | 9 | 1254 | ||
BC057098.1 | - | 9 | 1231 | 5715767 |