Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153065.3 | NP_694705.2 | 11 | 1250 | ||
GenBank mRNA | AK134274.1 | - | 11 | 1251 | ||
AK138442.1 | - | 11 | 1274 | |||
AK146469.1 | - | 11 | 1278 | |||
BC011321.1 | - | 10 | 1127 | 4007058 |