- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Amigo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004293.2 | NP_001004293.1 | 2 | 1103 | ||
NM_001287093.1 | NP_001274022.1 | 2 | 1103 | |||
NM_146137.3 | NP_666249.2 | 2 | 1138 | |||
GenBank mRNA | AB167509.1 | - | 1 | 759 | ||
AK035960.1 | - | 2 | 1034 | |||
AK173118.1 | - | 2 | 1092 | |||
AY237008.1 | - | 1 | 652 | |||
BC010598.1 | - | 2 | 1054 | 3498778 | ||
BC022907.1 | - | 2 | 1041 |