Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
PLEC
Gene Name:
plectin
Gene Aliases:
EBS1, EBSMD, EBSND, EBSO, EBSOG, EBSPA, HD1, LGMD2Q, PCN, PLEC1, PLEC1b, PLTN
Chromosome Location:
Chr.8: 143915147 - 143976800 on Build GRCh38
UniGene ID:
Hs.434248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10643
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000445.4
NP_000436.2
33
11000
NM_201378.3
NP_958780.1
32
10885
NM_201379.2
NP_958781.1
32
10953
NM_201380.3
NP_958782.1
32
11451
NM_201381.2
NP_958783.1
32
10848
NM_201382.3
NP_958784.1
32
10938
NM_201383.2
NP_958785.1
32
10891
NM_201384.2
NP_958786.1
32
10999
XM_005250976.3
XP_005251033.1
33
11493
XM_005250978.3
XP_005251035.1
33
11018
XM_005250979.4
XP_005251036.1
33
10977
XM_005250980.4
XP_005251037.1
33
11062
XM_005250981.3
XP_005251038.1
33
10899
XM_005250982.3
XP_005251039.1
33
10892
XM_005250983.2
XP_005251040.1
32
10816
XM_005250984.4
XP_005251041.1
33
10881
XM_006716588.3
XP_006716651.1
34
11070
XM_006716589.2
XP_006716652.1
33
10831
XM_006716590.3
XP_006716653.1
33
10941
XM_011517130.2
XP_011515432.1
33
11006
XM_011517131.2
XP_011515433.1
32
10925
XM_011517132.2
XP_011515434.1
32
7633
GenBank mRNA
AY480044.1
-
32
10957
AY480045.1
-
32
10828
AY480046.1
-
32
10953
AY480047.1
-
32
11451
AY480048.1
-
32
10848
AY480049.1
-
32
10877
AY480050.1
-
32
10891
AY480051.1
-
32
10999
BC007597.1
-
1
316
U53204.1
-
33
11002
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
hemidesmosome assembly
cell-cell adhesion
actin binding
protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0