Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052762.1 | - | 4 | 279 | ||
AK085717.1 | - | 4 | 370 | |||
AK154613.1 | - | 4 | 425 | |||
AK163171.1 | - | 4 | 372 | |||
AK170641.1 | - | 4 | 411 | |||
BB619031.1 | - | 4 | 472 | |||
BC005765.1 | - | 3 | 147 | |||
BC022976.1 | - | 4 | 357 | 5362925 | ||
BC024940.1 | - | 4 | 355 | |||
BC071258.1 | - | 4 | 290 |