Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207237.3 | NP_997120.1 | 11 | 2473 | ||
GenBank mRNA | AK048880.1 | - | 6 | 715 | ||
AK144486.1 | - | 10 | 2247 | |||
AK198104.1 | - | 2 | 68 | |||
BC030443.1 | - | 5 | 636 | |||
BC067023.1 | - | 12 | 2043 | 6848230 |