Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207237.3 | NP_997120.1 | 3 | 1470 | ||
GenBank mRNA | AK141122.1 | - | 3 | 948 | ||
AK144486.1 | - | 3 | 1439 | |||
BC067023.1 | - | 4 | 1040 | 6848230 |