Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153092.4 | NP_694732.3 | 7 | 1014 | ||
GenBank mRNA | AA867018.1 | - | 1 | 108 | ||
AB067574.1 | - | 6 | 862 | |||
AK077066.1 | - | 7 | 982 | |||
AK078478.1 | - | 7 | 1015 | |||
BC033270.1 | - | 7 | 956 | 3981211 |