Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172710.3 | NP_766298.2 | 20 | 3028 | ||
GenBank mRNA | AK030487.1 | - | 20 | 2893 | ||
AK122362.1 | - | 20 | 3028 | |||
BC026399.1 | - | 3 | 181 | |||
BC026655.1 | - | 8 | 751 | |||
BC094607.1 | - | 19 | 2573 | 6837048 | ||
BC117915.1 | - | 19 | 2529 | 40097358 | ||
BC117916.1 | - | 19 | 2531 | 40097359 |