Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115010.1 | NP_001108482.1 | 5 | 1379 | ||
NM_172714.4 | NP_766302.2 | 5 | 1247 | |||
GenBank mRNA | AK029288.1 | - | 6 | 693 | ||
AK030894.1 | - | 5 | 1381 | |||
AK134320.1 | - | 5 | 1245 | |||
AK220225.1 | - | 5 | 1242 | |||
BC055819.1 | - | 4 | 660 | |||
BC106126.1 | - | 4 | 883 | |||
BC125580.1 | - | 3 | 374 | |||
BC145025.1 | - | 3 | 374 | 9053573 |