Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcn1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172719.2 | NP_766307.2 | 9 | 853 | ||
GenBank mRNA | AK041717.1 | - | 9 | 842 | ||
AK090137.1 | - | 9 | 851 | |||
AK147257.1 | - | 9 | 846 | |||
AK154512.1 | - | 9 | 846 | |||
AK157964.1 | - | 6 | 4113 | |||
AK158040.1 | - | 9 | 844 | |||
BC150735.1 | - | 9 | 856 | 9087646 |