Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032242.3 | NP_115618.3 | 24 | 4535 | ||
XM_011512908.2 | XP_011511210.1 | 25 | 4637 | |||
XM_011512909.1 | XP_011511211.1 | 25 | 5230 | |||
GenBank mRNA | AK126101.1 | - | 2 | 1727 | ||
AK293425.1 | - | 5 | 654 | |||
X87832.2 | - | 24 | 4470 |