Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145148.2 | NP_660130.2 | 22 | 2879 | ||
GenBank mRNA | AB042027.1 | - | 22 | 2802 | ||
AF327856.1 | - | 21 | 2502 | |||
AF327857.1 | - | 20 | 2289 | |||
AK031491.1 | - | 11 | 2065 | |||
AK129262.1 | - | 21 | 2354 | |||
AK132280.1 | - | 22 | 2743 | |||
BC039783.1 | - | 1 | 309 | |||
BC052390.1 | - | 22 | 2748 | 5718942 |