Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182939.4 | NP_891984.1 | 4 | 560 | ||
GenBank mRNA | AK082539.1 | - | 4 | 561 | ||
AK084169.1 | - | 4 | 521 | |||
BC056340.1 | - | 4 | 380 | 5703151 | ||
BC062117.1 | - | 3 | 234 | |||
BC098464.1 | - | 4 | 326 | 30093511 | ||
BC110429.1 | - | 4 | 565 | 5350332 | ||
BC112399.1 | - | 3 | 138 |