Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181444.5 | NP_852109.2 | 3, 4 | 1109 | ||
GenBank mRNA | BC034063.1 | - | 2, 3 | 636 | ||
BC036173.1 | - | 3, 4 | 1133 | 4989149 | ||
BC036174.1 | - | 2, 3 | 913 | |||
BC039217.1 | - | 3, 4 | 1085 | 4205657 |
Set Membership: |
![]() |