Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301407.1 | NP_001288336.1 | 8 | 1007 | ||
NM_146173.3 | NP_666285.1 | 8 | 1010 | |||
GenBank mRNA | AF276890.1 | - | 8 | 1009 | ||
AK004954.1 | - | 8 | 983 | |||
AK172176.1 | - | 8 | 988 | |||
BC024685.1 | - | 8 | 968 | 3981492 |