Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000672.3 | NP_000663.1 | 5 | 505 | ||
NM_001102470.1 | NP_001095940.1 | 5 | 505 | |||
GenBank mRNA | AK092768.1 | - | 5 | 505 | ||
AK298290.1 | - | 1 | 74 | |||
BC039065.1 | - | 5 | 494 | 5184104 |