Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 6 | 689 | ||
NM_001322003.1 | NP_001308932.1 | 6 | 684 | |||
NM_001322004.1 | NP_001308933.1 | 5 | 549 | |||
NM_001322005.1 | NP_001308934.1 | 6 | 801 | |||
NM_001322006.1 | NP_001308935.1 | 6 | 689 | |||
NM_001322007.1 | NP_001308936.1 | 5 | 586 | |||
NM_001322008.1 | NP_001308937.1 | 4 | 446 | |||
NM_001322009.1 | NP_001308938.1 | 6 | 796 | |||
NM_001322010.1 | NP_001308939.1 | 5 | 549 | |||
NM_001322011.1 | NP_001308940.1 | 6 | 640 | |||
NM_001322012.1 | NP_001308941.1 | 6 | 635 | |||
NM_001322014.1 | NP_001308943.1 | 6 | 689 | |||
NM_001322015.1 | NP_001308944.1 | 6 | 859 | |||
NR_136154.1 | 6 | 689 | ||||
XM_017012342.1 | XP_016867831.1 | 6 | 614 | |||
GenBank mRNA | AB103082.1 | - | 6 | 602 | ||
AB103083.1 | - | 6 | 602 | |||
AB103086.1 | - | 3 | 284 | |||
AK294661.1 | - | 5 | 539 | |||
AK312390.1 | - | 6 | 638 | |||
BC008400.1 | - | 6 | 647 | |||
BC031832.1 | - | 6 | 653 | |||
BC093921.1 | - | 6 | 624 | 7939766 | ||
BP251389.1 | - | 2 | 159 | |||
CR981250.1 | - | 6 | 590 | |||
U14658.1 | - | 6 | 604 |