Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 11 | 1770 | ||
NM_001322003.1 | NP_001308932.1 | 11 | 1765 | |||
NM_001322004.1 | NP_001308933.1 | 10 | 1630 | |||
NM_001322005.1 | NP_001308934.1 | 11 | 1882 | |||
NM_001322006.1 | NP_001308935.1 | 10 | 1614 | |||
NM_001322007.1 | NP_001308936.1 | 10 | 1667 | |||
NM_001322008.1 | NP_001308937.1 | 9 | 1527 | |||
NM_001322009.1 | NP_001308938.1 | 11 | 1877 | |||
NM_001322010.1 | NP_001308939.1 | 9 | 1474 | |||
NM_001322011.1 | NP_001308940.1 | 11 | 1721 | |||
NM_001322012.1 | NP_001308941.1 | 11 | 1716 | |||
NM_001322013.1 | NP_001308942.1 | 10 | 1597 | |||
NM_001322014.1 | NP_001308943.1 | 11 | 1770 | |||
NM_001322015.1 | NP_001308944.1 | 11 | 1940 | |||
NR_136154.1 | 11 | 1770 | ||||
XM_006715744.3 | XP_006715807.1 | 4 | 846 | |||
XM_017012342.1 | XP_016867831.1 | 10 | 1595 | |||
GenBank mRNA | AB103086.1 | - | 8 | 1365 | ||
AK294661.1 | - | 10 | 1620 | |||
AK312390.1 | - | 11 | 1719 | |||
BC008400.1 | - | 11 | 1728 | |||
BC031832.1 | - | 11 | 1734 | |||
BC093921.1 | - | 11 | 1705 | 7939766 | ||
U14658.1 | - | 11 | 1685 |