Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146188.1 | NP_666300.1 | 5 | 950 | ||
GenBank mRNA | AK034731.1 | - | 6 | 1022 | ||
BC031749.1 | - | 5 | 950 | 4507203 |