Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205810.4 | NP_991379.3 | 2 | 925 | ||
GenBank mRNA | AK148044.1 | - | 2 | 921 | ||
AK148046.1 | - | 2 | 829 | |||
AK148052.1 | - | 2 | 829 | |||
AK148074.1 | - | 2 | 926 | |||
AK148108.1 | - | 2 | 830 | |||
AK148114.1 | - | 2 | 926 | |||
AK148166.1 | - | 2 | 830 | |||
AK148226.1 | - | 1 | 506 | |||
AK148241.1 | - | 2 | 830 | |||
AK148253.1 | - | 2 | 921 | |||
AK148256.1 | - | 2 | 921 | |||
AK148273.1 | - | 2 | 898 | |||
AK148279.1 | - | 2 | 879 | |||
AK148322.1 | - | 2 | 863 | |||
AK148388.1 | - | 2 | 830 | |||
BC116395.1 | - | 1 | 751 | |||
BC116396.1 | - | 1 | 746 |
Set Membership: |
![]() |