Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285997.1 | NP_001272926.1 | 4 | 454 | ||
NM_001285998.1 | NP_001272927.1 | 4 | 454 | |||
NM_145150.3 | NP_660132.2 | 4 | 454 | |||
GenBank mRNA | AK029140.1 | - | 4 | 441 | ||
AK081506.1 | - | 3 | 3627 | |||
AK088576.1 | - | 4 | 426 | |||
AK135778.1 | - | 2 | 79 | |||
BC005475.1 | - | 4 | 388 | 3592138 | ||
BC059808.1 | - | 4 | 399 | |||
CF736005.1 | - | 4 | 454 |