Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178707.4 | NP_848822.2 | 9 | 3410 | ||
GenBank mRNA | AK042675.1 | - | 9 | 3412 | ||
AK129089.1 | - | 9 | 3382 | |||
AK143618.1 | - | 9 | 3408 | |||
BC044728.2 | - | 7 | 1680 | |||
BC059073.1 | - | 7 | 2723 | |||
BC094321.1 | - | 7 | 3195 | 6401005 | ||
BC118972.1 | - | 8 | 3264 | 40098313 | ||
BC118973.1 | - | 8 | 3264 | 40098315 |