Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Picalm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252520.1 | NP_001239449.1 | 19 | 2172 | ||
NM_001252521.1 | NP_001239450.1 | 18 | 2163 | |||
NM_001252522.1 | NP_001239451.1 | 18 | 2037 | |||
NM_001252523.1 | NP_001239452.1 | 18 | 2022 | |||
NM_001252524.1 | NP_001239453.1 | 17 | 1998 | |||
NM_146194.4 | NP_666306.2 | 19 | 2187 | |||
GenBank mRNA | AK085472.2 | - | 14 | 1232 | ||
AK144960.1 | - | 19 | 1928 | |||
AK162360.1 | - | 19 | 1865 | |||
AK165066.1 | - | 13 | 1134 | |||
AK170373.1 | - | 17 | 2018 | |||
AK220291.1 | - | 19 | 1888 | |||
AY206701.1 | - | 19 | 1871 | |||
BC011470.1 | - | 19 | 1882 | 2651109 | ||
BC021491.1 | - | 17 | 1947 | 5345558 | ||
BC023843.1 | - | 18 | 1911 | 5346041 | ||
BC025566.1 | - | 18 | 2060 | 5322606 | ||
BC027116.1 | - | 15 | 1459 | |||
BC057683.1 | - | 18 | 1973 | 3671962 |