Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177977.1 | NP_001171448.1 | 11 | 1597 | ||
NM_001177978.1 | NP_001171449.1 | 11 | 1507 | |||
NM_146197.4 | NP_666309.1 | 11 | 1507 | |||
GenBank mRNA | AK057650.1 | - | 11 | 1507 | ||
AK140827.1 | - | 11 | 1598 | |||
AK143757.1 | - | 11 | 1503 | |||
AK165353.1 | - | 11 | 1505 | |||
AK165401.1 | - | 11 | 1496 | |||
BC024424.1 | - | 2 | 209 | |||
BC024450.1 | - | 1 | 101 | |||
BC031140.1 | - | 11 | 1483 | 4972673 |