Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145605.2 | NP_663580.2 | 5, 6 | 592 | ||
GenBank mRNA | AK089954.1 | - | 5, 6 | 555 | ||
AK161674.1 | - | 5, 6 | 568 | |||
BC012312.1 | - | 5, 6 | 592 | 2631632 | ||
BC023738.1 | - | 5, 6 | 563 | 5352770 | ||
BC058359.1 | - | 5, 6 | 540 | 6815650 |